siRNA Details
Gene Symbol: OARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145063.2 | NP_659500.1 | 3 | 308 | ||
GenBank mRNA | AJ420538.1 | - | 3 | 444 | ||
AK290939.1 | - | 3 | 311 | |||
AK310688.1 | - | 3 | 311 | |||
AK313361.1 | - | 3 | 349 | |||
BC011709.2 | - | 3 | 308 | 3942960 | ||
BC021579.1 | - | 3 | 371 |
