siRNA Details
Gene Symbol: ERH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004450.2 | NP_004441.1 | 3 | 217 | ||
GenBank mRNA | AK130850.1 | - | 3 | 203 | ||
AK312186.1 | - | 3 | 212 | |||
BC014301.2 | - | 3 | 200 | 3507241 | ||
BC071709.1 | - | 3 | 164 | 5503738 | ||
BT006877.1 | - | 2 | 146 | |||
CR450298.1 | - | 2 | 146 | |||
CR542178.1 | - | 2 | 146 | |||
D85758.1 | - | 3 | 220 | |||
U66871.1 | - | 3 | 217 |
