siRNA Details
Gene Symbol: FAM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145268.1 | NP_001138740.1 | 8 | 1537 | ||
NM_001145269.1 | NP_001138741.1 | 7 | 1186 | |||
NR_026879.1 | 8 | 1296 | ||||
GenBank mRNA | AK054745.1 | - | 8 | 1206 | ||
AK298753.1 | - | 7 | 1110 | |||
BC029175.1 | - | 8 | 1496 |
