siRNA Details
Gene Symbol: ATXN7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020725.1 | NP_065776.1 | 2, 3 | 278 | ||
NM_152749.3 | NP_689962.1 | 2, 3 | 278 | |||
XM_005250220.3 | XP_005250277.1 | 3, 4 | 361 | |||
XM_006715900.2 | XP_006715963.1 | 3, 4 | 361 | |||
XM_006715901.2 | XP_006715964.1 | 3, 4 | 361 | |||
XM_006715902.3 | XP_006715965.1 | 3, 4 | 361 | |||
XM_006715903.3 | XP_006715966.1 | 3, 4 | 361 | |||
XM_011515935.1 | XP_011514237.1 | 3, 4 | 361 | |||
XM_011515937.2 | XP_011514239.1 | 3, 4 | 361 | |||
XM_011515939.1 | XP_011514241.1 | 3, 4 | 361 | |||
XM_011515940.1 | XP_011514242.1 | 3, 4 | 361 | |||
XM_011515950.1 | XP_011514252.1 | 3, 4 | 361 | |||
XM_011515954.2 | XP_011514256.1 | 3, 4 | 361 | |||
XM_017011860.1 | XP_016867349.1 | 2, 3 | 193 | |||
GenBank mRNA | AB033044.1 | - | 2, 3 | 291 | ||
AK095475.1 | - | 2, 3 | 192 | |||
AK097574.1 | - | 2, 3 | 191 | |||
BC030616.2 | - | 2, 3 | 275 | 4796569 | ||
BQ431139.1 | - | 2, 3 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
