siRNA Details
Gene Symbol: CCDC129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095663.1 | - | 9 | 1384 | ||
AK128026.1 | - | 11 | 2025 | |||
AK295011.1 | - | 11 | 1566 | |||
AK304387.1 | - | 12 | 1623 | |||
BC132719.1 | - | 11 | 1514 | 40146741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
