siRNA Details
Gene Symbol: MS4A6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139249.2 | NP_640342.1 | 4 | 559 | ||
GenBank mRNA | AF333758.1 | - | 4 | 610 | ||
AF354931.1 | - | 4 | 559 | |||
AF354932.1 | - | 4 | 443 | |||
AF354933.1 | - | 4 | 499 | |||
BC034588.1 | - | 4 | 473 |
