siRNA Details
Gene Symbol: TXLNGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045128.1 | 2 | 285 | |||
NR_045129.1 | 2 | 285 | ||||
GenBank mRNA | AF332224.1 | - | 2 | 261 | ||
AK127387.1 | - | 2 | 381 | |||
AK303404.1 | - | 2 | 284 | |||
BC110817.1 | - | 3 | 400 | 6058816 | ||
BM263181.1 | - | 2 | 285 | |||
DA956406.1 | - | 2 | 384 |
