siRNA Details
Gene Symbol: NEIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135746.1 | NP_001129218.1 | 4, 5 | 828 | ||
NM_001135747.1 | NP_001129219.1 | 3, 4 | 688 | |||
NM_001135748.1 | NP_001129220.1 | 4, 5 | 642 | |||
NM_145043.2 | NP_659480.1 | 4, 5 | 1372 | |||
XM_005272382.2 | XP_005272439.1 | 4, 5 | 804 | |||
XM_005272383.2 | XP_005272440.1 | 3, 4 | 1100 | |||
XM_017013301.1 | XP_016868790.1 | 4, 5 | 1245 | |||
GenBank mRNA | AB079070.1 | - | 3, 4 | 697 | ||
AK056206.1 | - | 4, 5 | 1287 | |||
AK097389.1 | - | 3, 4 | 642 | |||
AK225837.1 | - | 4, 5 | 751 | |||
AK294224.1 | - | 4, 5 | 595 | |||
BC013952.2 | - | 4, 5 | 742 | 2964742 | ||
BC013964.2 | - | 4, 5 | 742 | 2964742 | ||
BX537529.1 | - | 5, 6 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
