siRNA Details
Gene Symbol: ETFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000126.3 | NP_000117.1 | 2 | 184 | ||
GenBank mRNA | AK292979.1 | - | 2 | 184 | ||
BC015526.1 | - | 2 | 128 | 3852346 | ||
BC029995.1 | - | 2 | 173 | |||
BC095457.1 | - | 2 | 109 | 30552923 | ||
BT009796.1 | - | 2 | 103 | |||
DA838974.1 | - | 2 | 184 | |||
J04058.1 | - | 2 | 103 | |||
KU178046.1 | - | 2 | 103 | |||
KU178047.1 | - | 3 | 236 | |||
KU178048.1 | - | 2 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
