siRNA Details
Gene Symbol: ANGPTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178127.4 | NP_835228.2 | 8 | 1367 | ||
XM_011542735.2 | XP_011541037.1 | 6 | 1166 | |||
XM_017017466.1 | XP_016872955.1 | 4 | 941 | |||
GenBank mRNA | AK291523.1 | - | 7 | 831 | ||
AY169281.1 | - | 7 | 825 | |||
AY358827.1 | - | 7 | 815 | |||
BC049170.1 | - | 8 | 1370 | 5285447 | ||
BC053957.1 | - | 8 | 1370 |
