siRNA Details
Gene Symbol: RNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098638.1 | NP_001092108.1 | 2 | 538 | ||
XM_011544889.2 | XP_011543191.1 | 2 | 613 | |||
GenBank mRNA | AB082522.1 | - | 2 | 538 |
