siRNA Details
Gene Symbol: RNF214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077239.1 | NP_001070707.1 | 5, 6 | 864 | ||
NM_001278249.1 | NP_001265178.1 | 5, 6 | 399 | |||
NM_207343.3 | NP_997226.2 | 5, 6 | 926 | |||
GenBank mRNA | AK298923.1 | - | 5, 6 | 858 | ||
AK300160.1 | - | 5, 6 | 399 | |||
BC146891.1 | - | 5, 6 | 864 | 9056819 | ||
BC146901.1 | - | 5, 6 | 864 | 9056829 |
