siRNA Details
Gene Symbol: RNF214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077239.1 | NP_001070707.1 | 7 | 1059 | ||
NM_001278249.1 | NP_001265178.1 | 7 | 594 | |||
NM_207343.3 | NP_997226.2 | 7 | 1121 | |||
GenBank mRNA | AK298923.1 | - | 7 | 1053 | ||
AK300160.1 | - | 7 | 594 | |||
BC031347.1 | - | 2 | 156 | |||
BC146891.1 | - | 7 | 1059 | 9056819 | ||
BC146901.1 | - | 7 | 1059 | 9056829 |
