siRNA Details
Gene Symbol: DSCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026719.2 | 3 | 1513 | |||
GenBank mRNA | AB066100.1 | - | 1 | 1280 | ||
BC029827.1 | - | 1 | 851 | |||
BC066653.1 | - | 3 | 1513 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DSCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026719.2 | 3 | 1513 | |||
GenBank mRNA | AB066100.1 | - | 1 | 1280 | ||
BC029827.1 | - | 1 | 851 | |||
BC066653.1 | - | 3 | 1513 |