siRNA Details
Gene Symbol: LINC01465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121682.1 | 1 | 1357 | |||
GenBank mRNA | AK098456.1 | - | 1 | 1358 | ||
BC038738.1 | - | 1 | 1060 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121682.1 | 1 | 1357 | |||
GenBank mRNA | AK098456.1 | - | 1 | 1358 | ||
BC038738.1 | - | 1 | 1060 |