siRNA Details
Gene Symbol: LINC00639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039982.1 | 3 | 341 | |||
GenBank mRNA | AK125018.1 | - | 3 | 336 | ||
BC035119.1 | - | 4 | 408 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039982.1 | 3 | 341 | |||
GenBank mRNA | AK125018.1 | - | 3 | 336 | ||
BC035119.1 | - | 4 | 408 |