siRNA Details
Gene Symbol: GOLGA6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093463.1 | - | 2 | 196 | ||
AK097414.1 | - | 2 | 196 | |||
BC129820.1 | - | 2 | 187 | 40113145 | ||
BC129821.1 | - | 2 | 187 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GOLGA6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093463.1 | - | 2 | 196 | ||
AK097414.1 | - | 2 | 196 | |||
BC129820.1 | - | 2 | 187 | 40113145 | ||
BC129821.1 | - | 2 | 187 |