siRNA Details
Gene Symbol: GOLGA6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304388.1 | NP_001291317.1 | 5 | 385 | ||
GenBank mRNA | AK093463.1 | - | 5 | 385 | ||
BC129820.1 | - | 5 | 376 | 40113145 | ||
BC129821.1 | - | 4 | 346 |
