siRNA Details
Gene Symbol: ZIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010879.3 | NP_001010879.2 | 4 | 1225 | ||
NM_001321145.1 | NP_001308074.1 | 3 | 1186 | |||
NM_001321146.1 | NP_001308075.1 | 3 | 1409 | |||
NM_001321147.1 | NP_001308076.1 | 2 | 1059 | |||
XM_011526761.1 | XP_011525063.1 | 4 | 1517 | |||
XM_011526762.1 | XP_011525064.1 | 4 | 1782 | |||
XM_011526763.1 | XP_011525065.1 | 3 | 1478 | |||
XM_011526764.1 | XP_011525066.1 | 4 | 860 | |||
XM_011526765.2 | XP_011525067.1 | 2 | 1616 | |||
XM_011526766.2 | XP_011525068.1 | 3 | 799 | |||
XM_011526767.2 | XP_011525069.1 | 2 | 1516 | |||
GenBank mRNA | AK092538.1 | - | 4 | 1059 | ||
AK128340.1 | - | 3 | 1022 | |||
BC103957.1 | - | 2 | 775 | 40011908 | ||
BC103958.1 | - | 4 | 941 | 40011910 | ||
BC103959.2 | - | 3 | 1125 | 40011911 |
