siRNA Details
Gene Symbol: TMEM145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173633.2 | NP_775904.2 | 7 | 615 | ||
XM_005258781.4 | XP_005258838.1 | 7 | 657 | |||
XM_011526791.2 | XP_011525093.1 | 7 | 609 | |||
XM_011526792.1 | XP_011525094.1 | 7 | 657 | |||
GenBank mRNA | AK075286.1 | - | 7 | 604 |
