siRNA Details
Gene Symbol: ACTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178525.4 | NP_848620.3 | 1 | 313 | ||
GenBank mRNA | AK292258.1 | - | 1 | 261 | ||
AY248901.1 | - | 1 | 188 | |||
BC024028.2 | - | 1 | 261 | 4824606 | ||
BC045752.1 | - | 1 | 305 | 4826735 | ||
HY011370.1 | - | 1 | 313 | |||
HY042660.1 | - | 1 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
