siRNA Details
Gene Symbol: MIR7-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027148.1 | 1 | 205 | |||
GenBank mRNA | AB058892.2 | - | 1 | 170 | ||
AB058893.2 | - | 1 | 170 | |||
BC090884.1 | - | 1 | 169 | |||
BC117294.1 | - | 1 | 143 | |||
BC117296.1 | - | 1 | 143 | |||
DB051621.1 | - | 1 | 205 |
