siRNA Details
Gene Symbol: C1orf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182581.3 | NP_872387.2 | 3 | 805 | ||
XM_005245103.3 | XP_005245160.1 | 2 | 799 | |||
XM_011509443.2 | XP_011507745.1 | 3 | 970 | |||
XM_011509444.1 | XP_011507746.1 | 3 | 817 | |||
GenBank mRNA | AY248900.1 | - | 3 | 785 | ||
BC032957.1 | - | 3 | 812 | 4827863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
