siRNA Details
Gene Symbol: LINC01114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038231.1 | 3 | 1873 | |||
GenBank mRNA | AK090794.1 | - | 2 | 1549 | ||
AK096621.1 | - | 3 | 1873 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038231.1 | 3 | 1873 | |||
GenBank mRNA | AK090794.1 | - | 2 | 1549 | ||
AK096621.1 | - | 3 | 1873 |