siRNA Details
Gene Symbol: RUFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198483.3 | NP_940885.2 | 8, 9 | 1122 | ||
NR_034176.1 | 9, 10 | 1983 | ||||
XM_011511014.2 | XP_011509316.1 | 10, 11 | 2064 | |||
XM_017003895.1 | XP_016859384.1 | 9, 10 | 2199 | |||
XM_017003896.1 | XP_016859385.1 | 9, 10 | 2196 | |||
XM_017003897.1 | XP_016859386.1 | 9, 10 | 2199 | |||
GenBank mRNA | AK128393.1 | - | 9, 10 | 1929 | ||
BC113712.1 | - | 2, 3 | 144 | 8322764 |
