siRNA Details
Gene Symbol: RUFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198483.3 | NP_940885.2 | 9 | 1286 | ||
NR_034176.1 | 10 | 2147 | ||||
XM_011511014.2 | XP_011509316.1 | 11 | 2228 | |||
XM_017003895.1 | XP_016859384.1 | 10 | 2363 | |||
XM_017003896.1 | XP_016859385.1 | 10 | 2360 | |||
XM_017003897.1 | XP_016859386.1 | 10 | 2363 | |||
GenBank mRNA | AK128393.1 | - | 10 | 2093 | ||
AK131246.1 | - | 8 | 1238 | |||
BC113712.1 | - | 3 | 308 | 8322764 |
