siRNA Details
Gene Symbol: ZNF619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145082.2 | NP_001138554.1 | 6 | 880 | ||
NM_001145083.1 | NP_001138555.1 | 4 | 577 | |||
NM_001145093.2 | NP_001138565.1 | 5 | 760 | |||
NR_026746.2 | 6 | 910 | ||||
XM_006713112.2 | XP_006713175.1 | 5 | 728 | |||
XM_006713113.3 | XP_006713176.1 | 4 | 584 | |||
XM_011533607.1 | XP_011531909.1 | 6 | 848 | |||
XM_011533608.1 | XP_011531910.1 | 6 | 802 | |||
XM_017006225.1 | XP_016861714.1 | 5 | 726 | |||
GenBank mRNA | AK075245.1 | - | 6 | 777 | ||
AK304139.1 | - | 4 | 577 | |||
AK304741.1 | - | 6 | 839 | |||
BC117169.1 | - | 4 | 512 | 40125720 | ||
BC143304.1 | - | 3 | 441 | |||
BC143306.1 | - | 4 | 512 | 9051811 | ||
BC143331.1 | - | 4 | 591 |
