siRNA Details
Gene Symbol: RNF212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131034.3 | NP_001124506.1 | 6, 7 | 510 | ||
NM_194439.4 | NP_919420.1 | 6, 7 | 510 | |||
XM_005272274.2 | XP_005272331.1 | 6, 7 | 508 | |||
XM_011513439.1 | XP_011511741.1 | 7, 8 | 699 | |||
XM_011513443.1 | XP_011511745.1 | 5, 6 | 678 | |||
XM_011513444.1 | XP_011511746.1 | 6, 7 | 508 | |||
XM_017008039.1 | XP_016863528.1 | 6, 7 | 536 | |||
XM_017008040.1 | XP_016863529.1 | 7, 8 | 574 | |||
GenBank mRNA | AK096160.1 | - | 6, 7 | 508 | ||
BC050356.1 | - | 6, 7 | 506 | 4815093 | ||
BC101258.1 | - | 6, 7 | 472 | 40023272 | ||
BC101259.1 | - | 6, 7 | 472 | 40023273 | ||
BC101260.1 | - | 6, 7 | 472 | 40023274 | ||
BG701603.1 | - | 6, 7 | 511 |
