siRNA Details
Gene Symbol: YIPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096895.1 | - | 4 | 575 | ||
BC103995.1 | - | 3 | 411 | |||
BC103996.1 | - | 4 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
