siRNA Details
Gene Symbol: RGMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012761.2 | NP_001012779.2 | 4 | 1157 | ||
XM_005271966.3 | XP_005272023.1 | 4 | 1013 | |||
XM_011543345.2 | XP_011541647.1 | 4 | 918 | |||
XM_011543346.2 | XP_011541648.1 | 2 | 1295 | |||
XM_011543347.2 | XP_011541649.2 | 3 | 841 | |||
XM_017009390.1 | XP_016864879.1 | 4 | 1405 | |||
XM_017009391.1 | XP_016864880.1 | 4 | 1303 | |||
GenBank mRNA | AK074887.1 | - | 4 | 1404 | ||
AK125284.1 | - | 1 | 56 | |||
BC067736.1 | - | 4 | 1157 | 30340500 |
