siRNA Details
Gene Symbol: PPIL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111298.2 | NP_001104768.2 | 2 | 797 | ||
NM_173672.4 | NP_775943.1 | 2 | 797 | |||
NR_104429.1 | 2 | 797 | ||||
XM_011535766.2 | XP_011534068.1 | 2 | 265 | |||
XM_011535767.2 | XP_011534069.1 | 2 | 265 | |||
XM_011535769.2 | XP_011534071.1 | 2 | 265 | |||
XM_017010774.1 | XP_016866263.1 | 2 | 265 | |||
GenBank mRNA | AK293674.1 | - | 2 | 249 | ||
AK315790.1 | - | 2 | 270 | |||
BC038716.1 | - | 2 | 256 | 5268756 | ||
DC419072.1 | - | 2 | 270 | |||
DQ363562.1 | - | 2 | 797 | |||
DQ423529.1 | - | 2 | 797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
