siRNA Details
Gene Symbol: ZNF707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095138.1 | - | 8 | 1186 | ||
BC014894.1 | - | 7 | 562 | 3910185 | ||
CV023324.1 | - | 3 | 281 | |||
DA188478.1 | - | 5 | 466 | |||
DA555314.1 | - | 6 | 534 | |||
DA917297.1 | - | 5 | 378 | |||
DA957023.1 | - | 6 | 489 | |||
DA990503.1 | - | 6 | 473 | |||
DB099015.1 | - | 6 | 534 | |||
DB106612.1 | - | 4 | 307 | |||
DB125792.1 | - | 4 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
