siRNA Details
Gene Symbol: CFAP157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012502.2 | NP_001012520.2 | 5 | 908 | ||
XM_006717064.3 | XP_006717127.2 | 5 | 1636 | |||
XM_011518559.2 | XP_011516861.2 | 5 | 1639 | |||
XM_017014629.1 | XP_016870118.1 | 5 | 1636 | |||
XM_017014630.1 | XP_016870119.1 | 5 | 975 | |||
GenBank mRNA | AK094948.1 | - | 5 | 832 | ||
AK126437.1 | - | 3 | 337 | |||
BC141809.1 | - | 5 | 881 | 40147564 |
