siRNA Details
Gene Symbol: GNPTAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024312.4 | NP_077288.2 | 18 | 3564 | ||
XM_006719593.2 | XP_006719656.1 | 18 | 3559 | |||
XM_011538731.2 | XP_011537033.1 | 18 | 3560 | |||
XM_017019961.1 | XP_016875450.1 | 18 | 3461 | |||
XM_017019962.1 | XP_016875451.1 | 18 | 3666 | |||
GenBank mRNA | AB033034.2 | - | 11 | 2455 | ||
AL359588.1 | - | 4 | 348 | |||
AM085438.1 | - | 18 | 3551 | |||
AY687932.1 | - | 18 | 3536 | |||
BC042615.1 | - | 6 | 1075 | |||
BC131787.1 | - | 18 | 3492 | 40126580 | ||
CR749871.1 | - | 4 | 348 |
