siRNA Details
Gene Symbol: GNPTAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024312.4 | NP_077288.2 | 10 | 1354 | ||
XM_006719593.2 | XP_006719656.1 | 10 | 1349 | |||
XM_011538731.2 | XP_011537033.1 | 10 | 1350 | |||
XM_017019961.1 | XP_016875450.1 | 10 | 1251 | |||
XM_017019962.1 | XP_016875451.1 | 10 | 1456 | |||
GenBank mRNA | AB033034.2 | - | 3 | 245 | ||
AK056137.1 | - | 10 | 1225 | |||
AM085438.1 | - | 10 | 1341 | |||
AY687932.1 | - | 10 | 1326 | |||
BC071687.1 | - | 10 | 1302 | |||
BC131787.1 | - | 10 | 1282 | 40126580 |
