siRNA Details
Gene Symbol: H2BFXP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003238.2 | 2 | 426 |
Gene Symbol: LOC100101478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130172.1 | 2 | 426 | |||
GenBank mRNA | AY730279.1 | - | 2 | 426 | ||
BC028039.1 | - | 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
