siRNA Details
Gene Symbol: CLEC4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080387.4 | NP_525126.2 | 5 | 606 | ||
XM_011520632.2 | XP_011518934.1 | 6 | 575 | |||
GenBank mRNA | AF411850.1 | - | 5 | 476 | ||
AK314071.1 | - | 5 | 593 | |||
AY115592.1 | - | 5 | 597 | |||
AY297446.1 | - | 5 | 605 | |||
AY486482.1 | - | 5 | 430 | |||
AY486483.1 | - | 5 | 430 | |||
BC032313.1 | - | 5 | 606 | 5218660 | ||
BI909298.1 | - | 5 | 555 |
