siRNA Details
Gene Symbol: OR5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003697.1 | NP_003688.1 | 1 | 459 | ||
GenBank mRNA | BC101239.2 | - | 1 | 461 | 40023020 | |
BC101241.1 | - | 1 | 459 | |||
BC101242.3 | - | 1 | 458 |
