siRNA Details
Gene Symbol: KRT79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175834.2 | NP_787028.1 | 1 | 316 | ||
GenBank mRNA | AJ564105.1 | - | 1 | 301 | ||
BC039148.1 | - | 1 | 318 | 4746574 | ||
BC063648.1 | - | 1 | 315 | 4804408 |
