siRNA Details
Gene Symbol: C17orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085430.2 | NP_001078899.2 | 7 | 1506 | ||
GenBank mRNA | BC041467.1 | - | 7 | 1506 | ||
BC093905.1 | - | 3 | 306 | 7939750 | ||
BC093907.1 | - | 3 | 306 | 7939752 |
