siRNA Details
Gene Symbol: C1orf174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207356.2 | NP_997239.2 | 2 | 166 | ||
XM_005244743.3 | XP_005244800.1 | 2 | 166 | |||
XM_011541323.2 | XP_011539625.1 | 2 | 2358 | |||
GenBank mRNA | AK125092.1 | - | 2 | 159 | ||
AK289493.1 | - | 2 | 166 | |||
BC035643.1 | - | 2 | 129 | |||
BC067302.1 | - | 2 | 158 | 30409047 |
