siRNA Details
Gene Symbol: CCDC190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005245125.3 | XP_005245182.1 | 2, 3 | 2987 | ||
XM_006711289.3 | XP_006711352.1 | 2, 3 | 469 | |||
GenBank mRNA | AK123573.1 | - | 2, 3 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
