siRNA Details
Gene Symbol: C2orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105519.2 | NP_001098989.1 | 3 | 419 | ||
NM_001322426.1 | NP_001309355.1 | 4 | 455 | |||
XM_011532810.2 | XP_011531112.1 | 5 | 622 | |||
XM_011532811.2 | XP_011531113.1 | 5 | 2118 | |||
XM_017003962.1 | XP_016859451.1 | 5 | 473 | |||
XM_017003963.1 | XP_016859452.1 | 3 | 417 | |||
XM_017003964.1 | XP_016859453.1 | 4 | 449 | |||
XM_017003965.1 | XP_016859454.1 | 5 | 473 | |||
XM_017003966.1 | XP_016859455.1 | 5 | 622 | |||
GenBank mRNA | AK091483.1 | - | 3 | 366 | ||
BX114846.1 | - | 4 | 428 | |||
HY021315.1 | - | 4 | 458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
