siRNA Details
Gene Symbol: ARSI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB448735.1 | - | 2 | 1211 | ||
AY875937.1 | - | 2 | 631 | |||
BC129995.1 | - | 2 | 688 | 40127932 | ||
BC129996.1 | - | 2 | 688 | 40127934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
