siRNA Details
Gene Symbol: ZNF680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178558.4 | NP_848653.2 | 4 | 1471 | ||
XM_006715959.2 | XP_006716022.1 | 5 | 1747 | |||
XM_017012126.1 | XP_016867615.1 | 7 | 2053 | |||
XM_017012127.1 | XP_016867616.1 | 6 | 1865 | |||
XM_017012128.1 | XP_016867617.1 | 6 | 1935 | |||
XM_017012129.1 | XP_016867618.1 | 5 | 1597 | |||
GenBank mRNA | AK074911.1 | - | 1 | 751 | ||
AK122937.1 | - | 4 | 1432 | |||
BC030700.1 | - | 4 | 1455 | 4837196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
