siRNA Details
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300932.1 | NP_001287861.1 | 4 | 625 | ||
NM_199136.4 | NP_954587.2 | 5 | 799 | |||
XM_011515369.2 | XP_011513671.1 | 5 | 794 | |||
XM_011515370.2 | XP_011513672.1 | 5 | 794 | |||
XM_011515372.2 | XP_011513674.1 | 5 | 939 | |||
GenBank mRNA | AL832127.1 | - | 6 | 3468 | ||
BC063130.1 | - | 5 | 713 | 30382529 | ||
BU958830.1 | - | 4 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
