siRNA Details
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127364.2 | NP_001120836.1 | 2 | 257 | ||
NM_199136.4 | NP_954587.2 | 2 | 257 | |||
XM_011515369.2 | XP_011513671.1 | 2 | 252 | |||
XM_011515370.2 | XP_011513672.1 | 2 | 252 | |||
XM_011515371.2 | XP_011513673.1 | 2 | 252 | |||
XM_011515372.2 | XP_011513674.1 | 2 | 397 | |||
XM_017012134.1 | XP_016867623.1 | 2 | 252 | |||
XM_017012135.1 | XP_016867624.1 | 2 | 397 | |||
GenBank mRNA | AK127774.1 | - | 2 | 204 | ||
AL832127.1 | - | 2 | 193 | |||
BC042034.1 | - | 2 | 239 | |||
BC063130.1 | - | 2 | 171 | 30382529 | ||
BI549491.1 | - | 2 | 244 | |||
CA867596.1 | - | 2 | 218 | |||
HY056255.1 | - | 2 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
