siRNA Details
Gene Symbol: ZNF517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317936.1 | NP_001304865.1 | 5 | 2138 | ||
NM_213605.2 | NP_998770.2 | 5 | 2159 | |||
XM_011517014.2 | XP_011515316.1 | 5 | 2698 | |||
XM_011517015.2 | XP_011515317.1 | 5 | 2140 | |||
XM_011517017.2 | XP_011515319.1 | 4 | 2557 | |||
XM_017013384.1 | XP_016868873.1 | 2 | 2083 | |||
GenBank mRNA | AK096527.1 | - | 6 | 2655 |
