siRNA Details
Gene Symbol: AKR1C8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027916.2 | 3 | 392 | |||
GenBank mRNA | AK131335.1 | - | 3 | 394 | ||
BC101204.1 | - | 3 | 386 | 40021857 | ||
BC101205.1 | - | 3 | 386 | 40021860 |
