siRNA Details
Gene Symbol: DCDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181807.3 | NP_861523.2 | 7, 8 | 1206 | ||
GenBank mRNA | AY247970.1 | - | 7, 8 | 1170 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DCDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181807.3 | NP_861523.2 | 7, 8 | 1206 | ||
GenBank mRNA | AY247970.1 | - | 7, 8 | 1170 |